Assay Details
| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 9 - 10 | 1739 | 103 | |
| NM_001270188.3 | NP_001257117.1 | 6 - 7 | 1396 | 103 | ||
More Information
Set Membership: |
|

pgp-5
Genomic Map