Assay Details
| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 11 - 12 | 1932 | 75 | |
| NM_001270188.3 | NP_001257117.1 | 8 - 9 | 1589 | 75 | ||
More Information
Set Membership: |
|

pgp-5
Genomic Map