Assay Details
| Gene Symbol: pgp-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077410.6 | NP_509811.2 | 11 - 12 | 2600 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgp-8| Gene Symbol: pgp-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077410.6 | NP_509811.2 | 11 - 12 | 2600 | 87 | |
Set Membership: |
|