Assay Details
| Gene Symbol: pgp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047822.6 | NP_001041287.2 | 5 - 6 | 833 | 136 | |
| NM_001047823.5 | NP_001041288.2 | 4 - 5 | 609 | 136 | ||
| NM_001270189.3 | NP_001257118.1 | 5 - 6 | 768 | 136 | ||
More Information
Set Membership: |
|

pgp-6
Genomic Map