Assay Details
| Gene Symbol: pxn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077433.5 | NP_509834.1 | 4 - 5 | 564 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pxn-2| Gene Symbol: pxn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077433.5 | NP_509834.1 | 4 - 5 | 564 | 79 | |
Set Membership: |
|