Assay Details
| Gene Symbol: asm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077493.7 | NP_509894.2 | 5 - 6 | 978 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

asm-2| Gene Symbol: asm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077493.7 | NP_509894.2 | 5 - 6 | 978 | 82 | |
Set Membership: |
|