Assay Details
| Gene Symbol: pgp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270213.3 | NP_001257142.1 | 14 - 15 | 3683 | 143 | |
| NM_001270214.5 | NP_001257143.1 | 13 - 14 | 3601 | 143 | ||
More Information
Set Membership: |
|

pgp-4
Genomic Map