Assay Details
| Gene Symbol: pgp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270213.3 | NP_001257142.1 | 3 - 4 | 239 | 78 | |
| NM_001270214.5 | NP_001257143.1 | 2 - 3 | 157 | 78 | ||
More Information
Set Membership: |
|

pgp-4
Genomic Map