Assay Details
| Gene Symbol: pgp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270213.3 | NP_001257142.1 | 8 - 9 | 1429 | 137 | |
| NM_001270214.5 | NP_001257143.1 | 7 - 8 | 1347 | 137 | ||
More Information
Set Membership: |
|

pgp-4
Genomic Map