Assay Details
| Gene Symbol: ttyh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383646.1 | 10 - 11 | 1660 | 84 | ||
| NM_077502.7 | NP_509903.1 | 9 - 10 | 1527 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ttyh-1| Gene Symbol: ttyh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383646.1 | 10 - 11 | 1660 | 84 | ||
| NM_077502.7 | NP_509903.1 | 9 - 10 | 1527 | 84 | ||
Set Membership: |
|