Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 2 - 3 | 677 | 82 | |
| NM_001270230.4 | NP_001257159.1 | 1 - 2 | 410 | 82 | ||
| NM_001307862.4 | NP_001294791.1 | 2 - 3 | 677 | 82 | ||
| NM_001307863.4 | NP_001294792.1 | 1 - 2 | 410 | 82 | ||
| NM_001373506.3 | NP_001360186.1 | 18 | 4283 | 82 | ||
| NM_001373507.3 | NP_001359540.1 | 19 | 4347 | 82 | ||
| NM_001383647.1 | 18 - 19 | 4360 | 82 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383647.1 transcripts.
Set Membership: |
|

hum-4
Genomic Map