Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 7 - 8 | 1244 | 77 | |
| NM_001270230.4 | NP_001257159.1 | 6 - 7 | 977 | 77 | ||
| NM_001270232.4 | NP_001257161.1 | 1 - 2 | 122 | 77 | ||
| NM_001307862.4 | NP_001294791.1 | 7 - 8 | 1244 | 77 | ||
| NM_001307863.4 | NP_001294792.1 | 6 - 7 | 977 | 77 | ||
| NM_001373506.3 | NP_001360186.1 | 23 | 4850 | 77 | ||
| NM_001373507.3 | NP_001359540.1 | 24 | 4914 | 77 | ||
| NM_001383647.1 | 23 - 24 | 4927 | 77 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map