Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 9 - 10 | 1534 | 85 | |
| NM_001270230.4 | NP_001257159.1 | 8 - 9 | 1267 | 85 | ||
| NM_001270232.4 | NP_001257161.1 | 3 - 4 | 412 | 85 | ||
| NM_001373507.3 | NP_001359540.1 | 26 | 5204 | 85 | ||
More Information
Set Membership: |
|

hum-4
Genomic Map