Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 10 - 11 | 1660 | 66 | |
| NM_001270230.4 | NP_001257159.1 | 9 - 10 | 1393 | 66 | ||
| NM_001270232.4 | NP_001257161.1 | 4 - 5 | 538 | 66 | ||
| NM_001307862.4 | NP_001294791.1 | 9 - 10 | 1549 | 66 | ||
| NM_001307863.4 | NP_001294792.1 | 8 - 9 | 1282 | 66 | ||
| NM_001373506.3 | NP_001360186.1 | 25 | 5155 | 66 | ||
| NM_001373507.3 | NP_001359540.1 | 27 | 5330 | 66 | ||
| NM_001383647.1 | 25 - 26 | 5232 | 66 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map