Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 12 - 13 | 2013 | 84 | |
| NM_001270230.4 | NP_001257159.1 | 11 - 12 | 1746 | 84 | ||
| NM_001270232.4 | NP_001257161.1 | 6 - 7 | 891 | 84 | ||
| NM_001307862.4 | NP_001294791.1 | 11 - 12 | 1902 | 84 | ||
| NM_001307863.4 | NP_001294792.1 | 10 - 11 | 1635 | 84 | ||
| NM_001373506.3 | NP_001360186.1 | 27 | 5508 | 84 | ||
| NM_001373507.3 | NP_001359540.1 | 29 | 5683 | 84 | ||
| NM_001383647.1 | 27 - 28 | 5585 | 84 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map