Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 15 - 16 | 2543 | 94 | |
| NM_001270230.4 | NP_001257159.1 | 14 - 15 | 2276 | 94 | ||
| NM_001270232.4 | NP_001257161.1 | 9 - 10 | 1421 | 94 | ||
| NM_001307862.4 | NP_001294791.1 | 14 - 15 | 2432 | 94 | ||
| NM_001307863.4 | NP_001294792.1 | 13 - 14 | 2165 | 94 | ||
| NM_001373506.3 | NP_001360186.1 | 30 | 6038 | 94 | ||
| NM_001373507.3 | NP_001359540.1 | 32 | 6213 | 94 | ||
| NM_001383647.1 | 30 - 31 | 6115 | 94 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map