Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 16 - 17 | 2654 | 114 | |
| NM_001270230.4 | NP_001257159.1 | 15 - 16 | 2387 | 114 | ||
| NM_001270232.4 | NP_001257161.1 | 10 - 11 | 1532 | 114 | ||
| NM_001307862.4 | NP_001294791.1 | 15 - 16 | 2543 | 114 | ||
| NM_001307863.4 | NP_001294792.1 | 14 - 15 | 2276 | 114 | ||
| NM_001373506.3 | NP_001360186.1 | 31 | 6149 | 114 | ||
| NM_001373507.3 | NP_001359540.1 | 33 | 6324 | 114 | ||
| NM_001383647.1 | 31 - 32 | 6226 | 114 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map