Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 21 - 22 | 3494 | 64 | |
| NM_001270230.4 | NP_001257159.1 | 20 - 21 | 3227 | 64 | ||
| NM_001270232.4 | NP_001257161.1 | 15 - 16 | 2372 | 64 | ||
| NM_001307862.4 | NP_001294791.1 | 20 - 21 | 3383 | 64 | ||
| NM_001307863.4 | NP_001294792.1 | 19 - 20 | 3116 | 64 | ||
| NM_001373506.3 | NP_001360186.1 | 36 | 6989 | 64 | ||
| NM_001373507.3 | NP_001359540.1 | 38 | 7164 | 64 | ||
| NM_001383647.1 | 36 - 37 | 7066 | 64 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map