Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270229.4 | NP_001257158.1 | 25 - 26 | 4044 | 90 | |
| NM_001270230.4 | NP_001257159.1 | 24 - 25 | 3777 | 90 | ||
| NM_001270232.4 | NP_001257161.1 | 19 - 20 | 2922 | 90 | ||
| NM_001307862.4 | NP_001294791.1 | 24 - 25 | 3933 | 90 | ||
| NM_001307863.4 | NP_001294792.1 | 23 - 24 | 3666 | 90 | ||
| NM_001373506.3 | NP_001360186.1 | 40 | 7539 | 90 | ||
| NM_001373507.3 | NP_001359540.1 | 42 | 7714 | 90 | ||
| NM_001383647.1 | 40 - 41 | 7616 | 90 | |||
More Information
Set Membership: |
|

hum-4
Genomic Map