Assay Details
| Gene Symbol: hum-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373506.3 | NP_001360186.1 | 3 | 260 | 159 | |
| NM_001373507.3 | NP_001359540.1 | 4 | 324 | 159 | ||
| NM_001383647.1 | 3 - 4 | 344 | 159 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383647.1 transcripts.
Set Membership: |
|

hum-4
Genomic Map