Assay Details
| Gene Symbol: sdc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077523.7 | NP_509924.1 | 10 - 11 | 4116 | 94 | |
| GenBank mRNA | AF111934.1 | - | 10 - 11 | 4110 | 94 | |
More Information
Set Membership: |
|

sdc-2
Genomic Map