Assay Details
| Gene Symbol: F52D10.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077536.6 | NP_509937.1 | 1 - 2 | 80 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F52D10.2| Gene Symbol: F52D10.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077536.6 | NP_509937.1 | 1 - 2 | 80 | 112 | |
Set Membership: |
|