Assay Details
| Gene Symbol: lyst-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077629.4 | NP_510030.2 | 18 - 19 | 3716 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lyst-1| Gene Symbol: lyst-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077629.4 | NP_510030.2 | 18 - 19 | 3716 | 59 | |
Set Membership: |
|