Assay Details
| Gene Symbol: lin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029416.4 | NP_001024587.1 | 21 - 22 | 2441 | 113 | |
| NM_001029417.4 | NP_001024588.1 | 11 - 12 | 1402 | 113 | ||
| GenBank mRNA | X92564.1 | - | 2425 | 113 | ||
| X92565.1 | - | 1402 | 113 | |||
More Information
Set Membership: |
|

lin-2
Genomic Map