Assay Details
| Gene Symbol: rgs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129662.4 | NP_001123134.1 | 3 - 4 | 457 | 112 | |
| NM_001270270.5 | NP_001257199.1 | 3 - 4 | 643 | 112 | ||
| NM_001383658.1 | 3 - 4 | 207 | 112 | |||
| NM_077723.6 | NP_510124.2 | 3 - 4 | 429 | 112 | ||
| GenBank mRNA | AF220265.1 | - | 3 - 4 | 392 | 112 | |
More Information
Set Membership: |
|

rgs-2
Genomic Map