Assay Details
| Gene Symbol: pgp-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077725.6 | NP_510126.1 | 12 - 13 | 3098 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgp-12| Gene Symbol: pgp-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077725.6 | NP_510126.1 | 12 - 13 | 3098 | 61 | |
Set Membership: |
|