Assay Details
| Gene Symbol: pgp-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373542.2 | NP_001359545.1 | 8 - 9 | 2271 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgp-13| Gene Symbol: pgp-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373542.2 | NP_001359545.1 | 8 - 9 | 2271 | 113 | |
Set Membership: |
|