Assay Details
| Gene Symbol: alh-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077731.7 | NP_510132.2 | 5 - 6 | 726 | 60 | |
| NM_077732.8 | NP_510133.2 | 5 - 6 | 718 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alh-13| Gene Symbol: alh-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077731.7 | NP_510132.2 | 5 - 6 | 726 | 60 | |
| NM_077732.8 | NP_510133.2 | 5 - 6 | 718 | 60 | ||
Set Membership: |
|