Assay Details
| Gene Symbol: alh-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077731.7 | NP_510132.2 | 6 - 7 | 1104 | 78 | |
| NM_077732.8 | NP_510133.2 | 6 - 7 | 1096 | 78 | ||
More Information
Set Membership: |
|

alh-13
Genomic Map