Assay Details
| Gene Symbol: gpc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383659.1 | 3 - 4 | 263 | 86 | ||
| GenBank mRNA | AF291848.1 | - | 1 - 2 | 84 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gpc-1| Gene Symbol: gpc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383659.1 | 3 - 4 | 263 | 86 | ||
| GenBank mRNA | AF291848.1 | - | 1 - 2 | 84 | 86 | |
Set Membership: |
|