Assay Details
| Gene Symbol: ppk-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077754.9 | NP_510155.3 | 9 - 10 | 2368 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ppk-3| Gene Symbol: ppk-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077754.9 | NP_510155.3 | 9 - 10 | 2368 | 73 | |
Set Membership: |
|