Assay Details
| Gene Symbol: scav-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077756.6 | NP_510157.2 | 9 - 10 | 1122 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scav-4| Gene Symbol: scav-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077756.6 | NP_510157.2 | 9 - 10 | 1122 | 133 | |
Set Membership: |
|