Assay Details
| Gene Symbol: snx-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083293.5 | NP_001076762.1 | 8 - 9 | 1048 | 102 | |
| NM_001083294.5 | NP_001076763.1 | 7 - 8 | 846 | 102 | ||
More Information
Set Membership: |
|

snx-17
Genomic Map