Assay Details
| Gene Symbol: spin-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077780.6 | NP_510181.3 | 6 - 7 | 1114 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spin-3| Gene Symbol: spin-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077780.6 | NP_510181.3 | 6 - 7 | 1114 | 139 | |
Set Membership: |
|