Assay Details
| Gene Symbol: skr-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077791.6 | NP_510192.1 | 1 | 464 | 92 | |
| GenBank mRNA | AF440519.1 | - | 1 | 478 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

skr-20| Gene Symbol: skr-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077791.6 | NP_510192.1 | 1 | 464 | 92 | |
| GenBank mRNA | AF440519.1 | - | 1 | 478 | 92 | |
Set Membership: |
|