Assay Details
| Gene Symbol: F46G10.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077820.6 | NP_510221.1 | 4 - 5 | 452 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F46G10.4| Gene Symbol: F46G10.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077820.6 | NP_510221.1 | 4 - 5 | 452 | 72 | |
Set Membership: |
|