Assay Details
| Gene Symbol: ptr-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270290.5 | NP_001257219.1 | 11 - 12 | 1922 | 148 | |
| NM_001270291.2 | NP_001257220.1 | 10 - 11 | 1700 | 148 | ||
More Information
Set Membership: |
|

ptr-24
Genomic Map