Assay Details
| Gene Symbol: ptr-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270290.5 | NP_001257219.1 | 16 - 17 | 2508 | 70 | |
| NM_001270291.2 | NP_001257220.1 | 15 - 16 | 2286 | 70 | ||
More Information
Set Membership: |
|

ptr-24
Genomic Map