Assay Details
| Gene Symbol: ptr-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270290.5 | NP_001257219.1 | 8 - 9 | 1425 | 129 | |
| NM_001270291.2 | NP_001257220.1 | 7 - 8 | 1203 | 129 | ||
More Information
Set Membership: |
|

ptr-24
Genomic Map