Assay Details
| Gene Symbol: sad-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083291.4 | NP_001076760.1 | 13 - 14 | 2519 | 77 | |
| NM_001083292.4 | NP_001076761.1 | 13 - 14 | 2525 | 77 | ||
| GenBank mRNA | AF316542.1 | - | 13 - 14 | 2507 | 77 | |
More Information
Set Membership: |
|

sad-1
Genomic Map