Assay Details
| Gene Symbol: cdk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077855.7 | NP_510256.1 | 2 - 3 | 309 | 72 | |
| GenBank mRNA | AF083878.1 | - | 1 - 2 | 122 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdk-4| Gene Symbol: cdk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077855.7 | NP_510256.1 | 2 - 3 | 309 | 72 | |
| GenBank mRNA | AF083878.1 | - | 1 - 2 | 122 | 72 | |
Set Membership: |
|