Assay Details
| Gene Symbol: cdk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077855.7 | NP_510256.1 | 4 - 5 | 802 | 106 | |
| GenBank mRNA | AF083878.1 | - | 3 - 4 | 615 | 106 | |
More Information
Set Membership: |
|

cdk-4
Genomic Map