Assay Details
| Gene Symbol: pab-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383665.1 | 3 - 4 | 354 | 79 | ||
| NM_077859.6 | NP_510260.1 | 2 - 3 | 609 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383665.1 transcripts.
Set Membership: |
|

pab-2
Genomic Map