Assay Details
| Gene Symbol: pab-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383665.1 | 4 - 5 | 667 | 81 | ||
| NM_077859.6 | NP_510260.1 | 3 - 4 | 922 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pab-2| Gene Symbol: pab-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383665.1 | 4 - 5 | 667 | 81 | ||
| NM_077859.6 | NP_510260.1 | 3 - 4 | 922 | 81 | ||
Set Membership: |
|