Assay Details
| Gene Symbol: pab-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383665.1 | 6 - 7 | 1671 | 97 | ||
| NM_077859.6 | NP_510260.1 | 5 - 6 | 1926 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pab-2| Gene Symbol: pab-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383665.1 | 6 - 7 | 1671 | 97 | ||
| NM_077859.6 | NP_510260.1 | 5 - 6 | 1926 | 97 | ||
Set Membership: |
|