Assay Details
| Gene Symbol: gck-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077889.4 | NP_510290.1 | 9 - 10 | 3365 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gck-4| Gene Symbol: gck-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077889.4 | NP_510290.1 | 9 - 10 | 3365 | 60 | |
Set Membership: |
|