Assay Details
| Gene Symbol: adm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368538.4 | NP_001355451.1 | 17 - 18 | 2635 | 113 | |
| NM_077890.6 | NP_510291.1 | 17 - 18 | 2635 | 113 | ||
More Information
Set Membership: |
|

adm-2
Genomic Map