Assay Details
| Gene Symbol: pix-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270298.4 | NP_001257227.1 | 3 - 4 | 254 | 71 | |
| NM_001270299.3 | NP_001257228.1 | 3 - 4 | 245 | 71 | ||
More Information
Set Membership: |
|

pix-1
Genomic Map