Assay Details
| Gene Symbol: alg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381140.1 | NP_001367206.1 | 5 - 6 | 2080 | 78 | |
| NM_001381141.1 | NP_001366765.1 | 5 - 6 | 2459 | 78 | ||
More Information
Set Membership: |
|

alg-1
Genomic Map