Assay Details
| Gene Symbol: tbc-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077935.7 | NP_510336.3 | 25 - 26 | 3691 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbc-19| Gene Symbol: tbc-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077935.7 | NP_510336.3 | 25 - 26 | 3691 | 124 | |
Set Membership: |
|